Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2E1 Mouse anti-Human, Clone: OTI5B9, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270531
Description
Cytochrome P450 2E1 Monoclonal antibody specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
Cytochrome P450 2E1 | |
OTI5B9 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase | |
Mouse | |
IgG1 | |
Immunogen affinity purified | |
Lipid and Metabolism | |
Primary | |
1571 | |
Human |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
LYOPH | |
Western Blot 1:500, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100 | |
Affinity Purified, Lyophilized | |
CYP2E1 | |
Full length human recombinant protein of human CYP2E1 (NP_000764) produced in HEK293T cell. | |
56.8 kDa | |
100 μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title