Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome p450 2J2 Mouse anti-Human, DyLight 755, Clone: OTI5C5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270539IR
Description
Cytochrome p450 2J2 Monoclonal antibody specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytochrome p450 2J2 | |
OTI5C5 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CYP2J2 | |
Full length recombinant protein of human Cytochrome p450 2J2 (NP_000766) produced in HEK293T cells. | |
Immunogen affinity purified | |
Lipid and Metabolism | |
Primary | |
1573.0 |
Immunohistochemistry, Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title