Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 4A1, 2C11 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen Cytochrome P450 4A1, 2C11
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125146
View Documents
Novus Biologicals
NBP31012225UL
25 μg
Each for $106.19
Add to cart
 
NB125145
View Documents
Novus Biologicals
NBP310122100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

Cytochrome P450 4A1, 2C11 Polyclonal antibody specifically detects Cytochrome P450 4A1, 2C11 in Rat samples. It is validated for Western Blot
Specifications

Specifications

Cytochrome P450 4A1, 2C11
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Rat
The immunogen is a synthetic peptide directed towards the c terminal region of rat Cytochrome P450 4A1, 2C110 (NP_787031.1). Peptide sequence PKVWPNPEVFDPSRFAPDSPRHSHSFLPFSGGARNCIGKQFAMSEMKVIV
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit