Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Enzyme CYP3A1 Mouse anti-Human, Mouse, Rat, Porcine, Clone: P6, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cytochrome P450 Enzyme CYP3A1
Clone P6
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122961
View Documents
Novus Biologicals
NBP30774410UG
100 μg
Each for $352.89
Add to cart
 
NB122962
View Documents
Novus Biologicals
NBP30774420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Cytochrome P450 Enzyme CYP3A1 Monoclonal antibody specifically detects Cytochrome P450 Enzyme CYP3A1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications

Specifications

Cytochrome P450 Enzyme CYP3A1
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
IgG1 Kappa
Primary
Store at 4°C.
P6
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Unconjugated
Mouse
Human, Mouse, Rat, Porcine
Full length protein (Rat). Rat liver cytochrome Cytochrome P450 Enzyme CYP3A1.
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit