Learn More
Cytochrome P450 26B1 Mouse anti-Human, Mouse, Rat, Janelia Fluor 549, Clone: T5P3G2*A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250195JF549
Catalog No. NBP50195549
Add to cart
Description
Description
CytochromeP45026B1 Monoclonal antibody specifically detects CytochromeP45026B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CytochromeP45026B1
T5P3G2&asterisk;A3
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CYP26B1
Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence)
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome
Mouse
IgG1 Kappa
0.1mL
Primary
56603