Learn More
CytochromeP4503A5 Mouse anti-Human, Janelia Fluor 646, Clone: F18P3B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250209JF646
Catalog No. NBP50209646
Add to cart
Description
Description
CytochromeP4503A5 Monoclonal antibody specifically detects CytochromeP4503A5 in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
CytochromeP4503A5
F18P3B6
Immunohistochemistry
Purified
CYP3A5
Ovalbumin-conjugated synthetic peptide DSRDGTLSGE (C-terminal sequence)
Protein G purified
Lipid and Metabolism
Primary
1577
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human