Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytohesin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody

$178.40 - $511.50

Specifications

Antigen Cytohesin 1
Concentration 0.77 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119069
View Documents
Invitrogen™
PA5119069
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5122049
View Documents
Invitrogen™
PA5122049
20 μL
Each for $178.40
Only null left
Add to Cart
 
Description

Description

Cytohesin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
Specifications

Specifications

Cytohesin 1
-20°C
Polyclonal
Liquid
IgG
Human
CYTH1
B2-1; CLM1; CTH-1; CYTH1; CYTIP; cytoadhesin 1; cytohesin 1; CYTOHESIN-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; SEC7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a
CYTH1
Primary
Antigen affinity chromatography
0.77 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
Q15438
9267
Recombinant protein Cytohesin 1. The antigen corresponds to amino acid range 1-398 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.