Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytohesin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100237
Description
Antibody detects endogenous levels of total Cytohesin2.
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Cytohesin 2 | |
Polyclonal | |
Unconjugated | |
CYTH2 | |
ARF exchange factor; ARF nucleotide binding site opener; ARF nucleotide-binding site opener; ARNO; CLM2; CTC-273B12.8; CTS18; CTS18.1; Cyth2; cytohesin 2; cytohesin-2; mSec7-2; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled-coil domains 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); Protein ARNO; PSCD2; PSCD2L; Sec7; SEC7 homolog B; Sec7b; SEC7L; Sec7p-L; Sec7p-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116692, 19158, 9266 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P63034, P63035, Q99418 | |
CYTH2 | |
A synthesized peptide derived from human CYTH2(Accession Q99418), corresponding to amino acid residues A215-W265. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction