Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytohesin 4 Monoclonal Antibody (OTI4A5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527436
Description
Cytohesin 4 Monoclonal antibody specifically detects Cytohesin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Cytohesin 4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UIA0 | |
CYTH4 | |
Full length human recombinant protein of CYTH4 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4A5 | |
Unconjugated | |
CYTH4 | |
CYT4; CYTH4; cytohesin 4; cytohesin-4; DJ63G5.1; PH, SEC7 and coiled-coil domain-containing protein 4; pleckstrin homology, Sec7 and coiled/coil domains 4; pleckstrin homology, Sec7 and coiled-coil domains 4; PSCD4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27128 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction