Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 1 Mouse anti-Human, Rat, Clone: LHK1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30777510UG

 View more versions of this product

Catalog No. NB123023


Add to cart

Description

Description

Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Rat samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin 1
LHK1
Unconjugated
10 mM PBS with 0.05% BSA
CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1
A 13-amino acid peptide (VRFVSTSYSGVTR) from the C-terminus of human Cytokeratin 1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human, Rat
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Purified
Mouse
IgG2a Kappa
100 μg
Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction
Primary
3848
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit