Learn More
Cytokeratin 10 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
$486.00 - $789.00
Specifications
Antigen | Cytokeratin 10 Ab-2 |
---|---|
Clone | DE-K10 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
MS611B1
|
Epredia
MS611B1 |
Concentrated | Biotin | 500 μL |
Each for $580.00
|
|
|||
MS611B
|
Epredia
MS611B |
Concentrated | Biotin | 1 mL |
Each for $789.00
|
|
|||
MS611P1ABX
|
Epredia
MS611P1ABX |
Concentrated | Unconjugated | 100 μL |
Each for $486.00
|
|
|||
MS611P1
|
Epredia
MS611P1 |
Concentrated | Unconjugated | 500 μL |
Each for $524.00
|
|
|||
MS611B0
|
Epredia
MS611B0 |
Concentrated | Biotin | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS611P0
|
Epredia
MS611P0 |
Concentrated | Unconjugated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS611PABX
|
Epredia
MS611PABX |
Concentrated | Unconjugated | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS611P
|
Epredia
MS611P |
Concentrated | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS611R1
|
Epredia
MS611R1 |
Prediluted | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS611R7
|
Epredia
MS611R7 |
Prediluted | Unconjugated | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
CK10 (Cytokeratin 10) is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.Host Species: Mouse
Clone: DE-K10
Isotype: IgG1
Species Reactivity: Human, Rat, Dog and Cat. Others not known.
Epitope: Not determined
Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium
Molecular Weight: 56.5 kDa
Positive Control: A431 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Cytokeratin 10 Ab-2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cytoskeletal Protein | |
Cytoskeletal preparation extracted from human ectocervical epithelium | |
Primary |
DE-K10 | |
Monoclonal | |
IVD | |
Canine, Feline, Human, Rat | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.