Learn More
Cytokeratin 10 Mouse anti-Human, Canine, Feline, Alexa Fluor™ 750, Clone: SPM623, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP254332AF750
Catalog No. NP24332A70
Add to cart
Description
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human,Canine,Feline samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
Cytokeratin 10
SPM623
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human, Canine, Feline
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
KRT10
Cytoskeletal preparation extracted from human ectocervical epithelium
0.1 mL
Primary
3858