Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 10 Rabbit anti-Human, Canine, Feline, Clone: KRT10/1948R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | Cytokeratin 10 |
---|---|
Clone | KRT10/1948R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123130
|
Novus Biologicals
NBP30782820UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Canine, Feline samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cytokeratin 10 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
3858 | |
IgG | |
Protein A or G purified |
KRT10/1948R | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Canine, Feline | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
Recombinant human full-length Cytokeratin 10 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title