Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 13 Mouse anti-Human, Mouse, Rat, Rabbit, Clone: KRT13/2850, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30784810UG

 View more versions of this product

Catalog No. NB123169


Add to cart

Description

Description

Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin 13
KRT13/2850
Unconjugated
10 mM PBS with 0.05% BSA
CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781
esophageal keratins of rabbit origin
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human, Mouse, Rat, Rabbit
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Cell Biology, Cellular Markers
Primary
3860
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit