Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14 Mouse anti-Human, Clone: KRT14/4132, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30784910UG

 View more versions of this product

Catalog No. NB123171


Add to cart

Description

Description

Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Cytokeratin 14
KRT14/4132
Unconjugated
10 mM PBS with 0.05% BSA
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
Recombinant fragment (around aa351-472) of human Cytokeratin 14 protein (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2c Kappa
100 μg
Cell Biology, Cellular Markers
Primary
3861
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit