Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 14, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9020P1

 View more versions of this product

Catalog No. RB9020P1


Add to cart

Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Cytokeratin 14
Polyclonal
Unconjugated
Rabbit
IgG
IVD
Primary
Polyclonal
Western Blot
200µg/mL
Concentrated
A synthetic peptide derived from C-terminal of human Keratin 14
500μL
Cytoskeletal Protein
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit