Learn More
Epredia™ Lab Vision™ Cytokeratin 15 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunofluorescence applications with Epredia™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1068P1ABX
Description
Keratin 15 is a type I keratin without a defined type II partner. Keratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail. Expression of keratin 15 is downregulated in some hyperproliferating situations, such as psoriasis and hypertrophic scars. Because keratinocytes in psoriasis and hypertrophic scars are activated, it is suggested that keratin 15 expression is not compatible with keratinocyte activation and the keratin 15 gene is downregulated to maintain the activated phenotype.Host Species: Mouse
Clone: LHK15
Isotype: IgG2a/κ
Species Reactivity: Human, Cow, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Cytokeratin 15 Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cytoskeletal Protein | |
Bovine, Human, Mouse, Rat | |
IgG2a κ |
Immunofluorescence | |
LHK15 | |
Unconjugated | |
A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15 | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.