Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 15 Mouse anti-Human, Clone: KRT15/2958, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cytokeratin 15
Clone KRT15/2958
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122702
View Documents
Novus Biologicals
NBP30761420UG
20 μg
Each for $138.37
Add to cart
 
NB122701
View Documents
Novus Biologicals
NBP30761410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Cytokeratin 15
0.2 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
Recombinant full-length human Cytokeratin 15 protein
Monoclonal
Protein A or G purified
KRT15/2958
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
3866
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit