Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 15 Mouse anti-Human, Rat, Bovine, PE-Cyanine7, Clone: SPM190, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254453PECY7

Catalog No. N254453PEC7


Add to cart

Description

Description

Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytokeratin 15
SPM190
Flow Cytometry
Conjugated, Purified
KRT15
A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15
Protein A or G purified
Cytoskeleton Markers
Primary
3866.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Bovine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit