Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 15 Rabbit anti-Human, Rat, Bovine, Clone: KRT15/2103R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | Cytokeratin 15 |
---|---|
Clone | KRT15/2103R |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122716
|
Novus Biologicals
NBP30762120UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 15 | |
0.2 mg/ml | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
3866 | |
IgG | |
Protein A purified |
KRT15/2103R | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Bovine | |
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
Recombinant human full-length Cytokeratin 15 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title