Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 16 Mouse anti-Human, Clone: KRT16/1714, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cytokeratin 16
Clone KRT16/1714
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122769
View Documents
Novus Biologicals
NBP30764810UG
100 μg
Each for $352.89
Add to cart
 
NB122770
View Documents
Novus Biologicals
NBP30764820UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Cytokeratin 16 Monoclonal antibody specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Cytokeratin 16
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK
Recombinant fragment from the C-terminal of human Cytokeratin 16
Monoclonal
Protein A or G purified
KRT16/1714
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Unconjugated
Mouse
Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
3868
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit