Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 16 Mouse anti-Human, Clone: rKRT16/1714, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Cytokeratin 16 |
---|---|
Clone | rKRT16/1714 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122834
|
Novus Biologicals
NBP30768020UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cytokeratin 16 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
3868 | |
IgG1 κ | |
Protein A or G purified |
rKRT16/1714 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK | |
Recombinant fragment from the C-terminal of human Cytokeratin 16 | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title