Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 16 Rabbit anti-Human, Clone: KRT16/2043R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$301.00

Specifications

Antigen Cytokeratin 16
Clone KRT16/2043R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122836
SDP
View Documents
Novus Biologicals
NBP30768120UG
20 μg
Each of 1 for $301.00
Add to Cart
 
Description

Description

Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin 16
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
3868
IgG
Protein A or G purified
KRT16/2043R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK
Recombinant fragment from the C-terminal of human Cytokeratin 16
Primary
Store at 4°C.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit