Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 17 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 17 Ab-1, Mouse Monoclonal Antibody.

$149.00 - $571.00

Specifications

Antigen Cytokeratin 17 Ab-1
Clone E3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS489R7
View Documents
Thermo Scientific™
MS489R7
7mL
Each for $282.00
Add to cart
 
MS489S
View Documents
Thermo Scientific™
MS489S
1mL
Each for $571.00
Add to cart
 
MS489S0
View Documents
Thermo Scientific™
MS489S0
100μL
Each for $149.00
Add to cart
 
MS489S1
View Documents
Thermo Scientific™
MS489S1
500μL
Each for $405.00
Add to cart
 
MS489R1
View Documents
Thermo Scientific™
MS489R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 17 Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

The type I keratin 17 (K17) shows a peculiar localization in human epithelial appendages including hair follicles, which undergo a growth cycle throughout adult life. Additionally K17 is induced, along with K6 and K16, early after acute injury to human skin. Predominant expression of K17 and the frequent expression of K8 and K19, with little K6/K16 and K1/K10 expression are the characteristic features of basal cell carcinomas (BCC), suggesting that BCC is differentiated towards undifferentiated follicular epithelia, most probably hair bulge cells.

Host Species: Mouse

Clone: E3

Isotype: IgG2b/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Cytokeratin preparation from rat enterocytes

Molecular Weight: 46 kDa

Positive Control: Squamous cell CA, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Specifications

Cytokeratin 17 Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2b kappa
E3
Monoclonal
Murine
Cytoskeletal Protein
Keratin preparation from rat enterocytes
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit