Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 17 Mouse anti-Human, Alexa Fluor™ 350, Clone: 9F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242619AF350

Catalog No. NP242619A30


Add to cart

Description

Description

Cytokeratin 17 Monoclonal specifically detects Cytokeratin 17 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Cytokeratin 17
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
KRT17
Recombinant human KRT17(1-432aa) purified from E. coli
100 μL
Primary
Human
IgG2a κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
9F3
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Mouse
Protein A purified
Cancer
3872.0
Store at 4°C in the dark.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit