Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 17 Mouse anti-Human, Clone: KRT17/4604, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cytokeratin 17
Clone KRT17/4604
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122837
View Documents
Novus Biologicals
NBP30768210UG
100 μg
Each for $352.89
Add to cart
 
NB122838
View Documents
Novus Biologicals
NBP30768220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Cytokeratin 17
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Recombinant full-length human Cytokeratin 17 protein
Monoclonal
Protein A or G purified
KRT17/4604
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
3872
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit