Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 17 Mouse, Clone: SPM560, Novus Biologicals™
Mouse Monoclonal Antibody
$229.00 - $459.00
Specifications
Antigen | Cytokeratin 17 |
---|---|
Clone | SPM560 |
Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234378F
|
Novus Biologicals
NBP2343780.02MG |
0.02 mg |
Each for $229.00
|
|
NBP234378A
|
Novus Biologicals
NBP2343780.1MG |
0.1 mg |
Each for $459.00
|
|
NBP234378B
|
Novus Biologicals
NBP2343780.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Cytokeratin 17 Monoclonal specifically detects Cytokeratin 17 in Human, Rat, Porcine, Bovine, Goat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 17 | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Porcine, Bovine, Goat | |
Q04695 | |
3872 | |
The cytoskeletal fraction of rat colon epithelium | |
Primary | |
Store at 4C. |
SPM560 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
KRT17 | |
IgG2b κ | |
Protein A purified | |
46 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title