Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 18 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

$190.00 - $533.00

Specifications

Antigen Cytokeratin 18 Ab-1
Clone DC10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS142B
View Documents
Thermo Scientific™
MS142B
Biotinylated Ab with BSA and Azide 1mL
Each for $533.00
Add to cart
 
MS142B0
View Documents
Thermo Scientific™
MS142B0
Biotinylated Ab with BSA and Azide 100μL
Each for $197.10
Add to cart
 
MS142B1
View Documents
Thermo Scientific™
MS142B1
Biotinylated Ab with BSA and Azide 500μL
Each for $384.00
Add to cart
 
MS142P
View Documents
Thermo Scientific™
MS142P
Purified Ab with BSA and Azide 1mL
Each for $358.00
Add to cart
 
MS142P0
View Documents
Thermo Scientific™
MS142P0
Purified Ab with BSA and Azide 100μL
Each for $190.00
Add to cart
 
MS142P1
View Documents
Thermo Scientific™
MS142P1
Purified Ab with BSA and Azide 500μL
Each for $303.00
Add to cart
 
MS142P1ABX
View Documents
Thermo Scientific™
MS142P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $316.00
Add to cart
 
MS142PABX
View Documents
Thermo Scientific™
MS142PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $409.50
Add to cart
 
MS142R7
View Documents
Thermo Scientific™
MS142R7
Ready-to-use for IHC (UVLP) 7mL
Each for $281.50
Add to cart
 
MS142R1
View Documents
Thermo Scientific™
MS142R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 18 Ab-1, Mouse Monoclonal Antibody.

Keratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with keratin 8. It was reported that tissues from gastrointestinal tract are positive for both keratin 8 and 18 but do not contain keratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for keratin 18.

Host Species: Mouse

Clone: DC10

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Human breast cancer PMC 42 cells

Molecular Weight: 45 kDa

Positive Control: HT29 cells, Skin, Breast or Lung CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Specifications

Specifications

Cytokeratin 18 Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Cytoskeletal Protein
Human breast cancer PMC 42 cells
DC10
Monoclonal
IVD
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit