Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Antibody (KRT18/835) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2479860.2MG
Description
Cytokeratin 18 Monoclonal antibody specifically detects Antigen in Bovine (Negative), Canine (Negative), Hamster (Negative), Human, Mouse (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Cytokeratin 18 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Mouse | |
45 kDa | |
0.2 mg | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers | |
3875 | |
Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative) | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
KRT18/835 | |
Unconjugated | |
PBS with No Preservative | |
KRT18 | |
Recombinant full-length human KRT18 protein | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It is reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction