Learn More
Cytokeratin 18 Mouse anti-Human, Bovine, Canine, Hamster, Mouse, Porcine, Rat, Sheep, Alexa Fluor™ 750, Clone: DC10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233082AF750
Catalog No. NP233082A70
Add to cart
Description
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human,Bovine,Canine,Hamster,Mouse,Porcine,Rat,Sheep samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Cytokeratin 18
DC10
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
KRT18
Human breast cancer PMC 42 cells were used as immunogen to generate the Keratin 18 antibody.
0.1 mL
Primary
3875
Bovine, Canine, Hamster, Human, Mouse, Porcine, Rat, Sheep