Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 18 Mouse anti-Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative), PE/Atto594, Clone: DC10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP233082PEATT594
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry.Specifications
Cytokeratin 18 | |
DC10 | |
Flow Cytometry | |
Conjugated, Purified | |
KRT18 | |
Human breast cancer PMC 42 cells were used as immunogen to generate the Keratin 18 antibody. | |
Protein G purified | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
Primary | |
3875.0 | |
Bovine (Negative), Canine (Negative), Hamster (Negative), Human, Mouse (Negative), Porcine (Negative), Rat (Negative) |
Flow Cytometry | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title