Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 19 Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with  Thermo Scientific™ Cytokeratin 19 Ab-4, Mouse Monoclonal Antibody.

$151.00 - $641.50

Specifications

Antigen Cytokeratin 19 Ab-4
Clone BA17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1671P
View Documents
Thermo Scientific™
MS1671P
Purified Ab with BSA and Azide 1mL
Each for $558.00
Add to cart
 
MS1671P0
View Documents
Thermo Scientific™
MS1671P0
Purified Ab with BSA and Azide 100μL
Each for $151.00
Add to cart
 
MS1671P1
View Documents
Thermo Scientific™
MS1671P1
Purified Ab with BSA and Azide 500μL
Each for $437.50
Add to cart
 
MS1671P1ABX
View Documents
Thermo Scientific™
MS1671P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $482.00
Add to cart
 
MS1671PABX
View Documents
Thermo Scientific™
MS1671PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $641.50
Add to cart
 
MS1671R7
View Documents
Thermo Scientific™
MS1671R7
Ready-to-use for IHC (UVLP) 7mL
Each for $222.00
Add to cart
 
MS1671R1
View Documents
Thermo Scientific™
MS1671R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with  Thermo Scientific™ Cytokeratin 19 Ab-4, Mouse Monoclonal Antibody.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: BA17

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Detergent soluble extract of human mammary epithelium

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin 19 Ab-4
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG2a kappa
BA17
Monoclonal
Murine
Cytoskeletal Protein
Detergent soluble extract of human mammary epithelium
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit