Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Mouse anti-Human, Clone: 963410, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals MAB35062
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytokeratin 19 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
KRT19 | |
E.coli-derived recombinant human Cytokeratin 19 Gln311-Lys370 Accession # P08727 | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
3880 | |
Purified |
Western Blot, Immunohistochemistry | |
963410 | |
Western Blot 0.25 - 1.0 ug/ml, Immunohistochemistry 5 ug/ml | |
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only