Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 19, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Manufacturer:  Thermo Scientific™ RB9021R1

 View more versions of this product

Catalog No. RB9021R1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 19, Rabbit Polyclonal Antibody.

Keratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 19

Molecular Weight: 40 kDa

Positive Control: LS174T

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin 19
Polyclonal
Unconjugated
Prediluted
A synthetic peptide derived from C-terminal of human Keratin 19
1mL
Cytoskeletal Protein
Primary
Human
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Ready-to-use for IHC (UVLP)
Rabbit
IgG
RUO
Primary
Polyclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit