Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Mouse anti-Human, Clone: KRT20/1991, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30728510UG

 View more versions of this product

Catalog No. NB122043


Add to cart

Description

Description

Cytokeratin 20 Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Cytokeratin 20
KRT20/1991
Unconjugated
10 mM PBS with 0.05% BSA
CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT
Recombinant fragment of human Cytokeratin 20 protein (around aa 196-323) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1 Kappa
100 μg
Cell Biology, Cellular Markers
Primary
54474
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit