Learn More
Cytokeratin 20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 173291AP
Catalog No. 50-173-1513
Add to cart
Description
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
Specifications
Cytokeratin 20
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25030, P35900, Q9D312
CK 20, CK20, Cytokeratin 20, K20, keratin 20, KRT20, KRT21, Protein IT
Rabbit
IgG
150 μL
-20°C
Primary
286912, 54474, 66809
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KRT20
Liquid
Krt20
Cytokeratin 20 Fusion Protein Ag11308
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat