Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3 Antibody (KRT3/2130), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279852UV
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 3 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT3 | |
Recombinant full-length human Cytokeratin 3 protein (Uniprot: P12035) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
KRT3/2130 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG1 κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction