Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3, Mouse anti-Human, Alexa Fluor 532, Clone: KRT3/2130, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279852AF532
Description
Cytokeratin 3 Monoclonal antibody specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Cytokeratin 3 | |
KRT3/2130 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
KRT3 | |
Recombinant full-length human Cytokeratin 3 protein | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title