Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 3, Mouse anti-Human, Clone: KRT3/2130, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | Cytokeratin 3 |
---|---|
Clone | KRT3/2130 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Cytokeratin 3 | |
0.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
P12035 | |
3850 | |
Recombinant full-length human Cytokeratin 3 protein | |
Primary | |
Store at 4°C. |
KRT3/2130 | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
KRT3 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title