Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 5/14 Monoclonal Antibody (16.4)
GREENER_CHOICE

Catalog No. 01674358
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-358 Supplier Invitrogen™ Supplier No. MA112590
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/14 Monoclonal Antibody for Western Blot, IHC (F)

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 5/14
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 16.4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene KRT5
Gene Accession No. P02533, P13647
Gene Alias 58 kda cytokeratin; CK14; CK-14; CK5; CK-5; cytokeratin 14; cytokeratin-14; Cytokeratin-5; DDD; DDD1; EBS2; EBS3; EBS4; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K14; K5; keratin 14; keratin 14, type I; keratin 5; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin, type I cytoskeletal 14; Keratin, type II cytoskeletal 5; keratin-14; Keratin-5; KRT14; KRT5; KRT5A; NFJ; Type-II keratin Kb5
Gene Symbols KRT14, KRT5
Host Species Mouse
Immunogen Cytokeratin enriched extract of feline tongue epithelium.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101090912, 101100735, 3852, 3861
Target Species Human, Feline
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.