Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5/8 Mouse, Clone: SPM268, Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$574.00
Specifications
Antigen | Cytokeratin 5/8 |
---|---|
Clone | SPM268 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot : 0.5 - 1.0 ug/ml, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247823A
|
Novus Biologicals
NBP2478230.1MG |
0.1 mg |
Each for $574.00
|
|
NBP247823B
|
Novus Biologicals
NBP2478230.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Cytokeratin 5/8 Monoclonal specifically detects Cytokeratin 5/8 in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Ferret, Sheep, Chicken (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
Cytokeratin 5/8 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Porcine, Canine, Ferret, Hamster, Primate, Sheep, Chicken (Negative), Xenopus (Negative) | |
cell proliferation-inducing protein 46, CK18, CK5, CYK18, DDD, K18, K1CO, K5, Keratin, Keratin 18, Keratin 5, KRT18, krt5, KRT5A, KRTB, type I cytoskeletal 18, type II cytoskeletal 5 | |
KRT5 | |
IgG1 κ | |
Protein A or G purified | |
It reacts with keratin 5 (58kDa) and keratin 8 (52.5kDa). Simple epithelia express cytokeratin 8 in combination with 18. On the other hand, basal cells of stratified epithelia express cytokeratin 5 paired with 14. This antibody therefore, reacts with a wide range of epithelia and their carcinomas. |
SPM268 | |
Western Blot : 0.5 - 1.0 ug/ml, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
3852 | |
Cytoskeletal preparation from HeLa cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title