Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody.

$161.00 - $425.50

Specifications

Antigen Cytokeratin 5 Ab-1
Clone XM26
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1896R7
View Documents
Thermo Scientific™
MS1896R7
7mL
Each for $383.50
Add to cart
 
MS1896S0
View Documents
Thermo Scientific™
MS1896S0
100μL
Each for $161.00
Add to cart
 
MS1896S1
View Documents
Thermo Scientific™
MS1896S1
500μL
Each for $425.50
Add to cart
 
MS1896R1
View Documents
Thermo Scientific™
MS1896R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS1896S
View Documents
Thermo Scientific™
MS1896S
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification. Point mutations in cytokeratin5 gene can cause various types of epidermolysis bullosa simplex. It is expressed in most epithelial and biphasic mesotheliomas.

Host Species: Mouse

Clone: XM26

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5

Molecular Weight: 58 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Cytokeratin 5 Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a kappa
XM26
Monoclonal
Murine
Cytoskeletal Protein
Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit