Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 6 Mouse anti-Human, Mouse, Clone: KRT6B/2116, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Cytokeratin 6
Clone KRT6B/2116
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123070
View Documents
Novus Biologicals
NBP30779820UG
20 μg
Each for $138.37
Add to cart
 
NB123069
View Documents
Novus Biologicals
NBP30779810UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Cytokeratin 6 Monoclonal antibody specifically detects Cytokeratin 6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Cytokeratin 6
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse
Allergen Hom s 5, CK6A, CK-6A, CK6C, CK6D, CK-6D, cytokeratin 6A, cytokeratin 6C, cytokeratin 6D, Cytokeratin-6A, Cytokeratin-6D, K6A, K6C, K6D, keratin 6A, keratin 6C, keratin 6D, keratin, epidermal type II, K6A, keratin, type II cytoskeletal 6A, Keratin-6A, KRT6C, KRT6D, Type-II keratin Kb6
Recombinant full-length human Cytokeratin 6 protein
Monoclonal
Protein A or G purified
KRT6B/2116
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cell Biology, Cellular Markers, Stem Cell Markers
10 mM PBS with 0.05% BSA
3853
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit