Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.
Supplier: Epredia MS1352PABX
Description
Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).Host Species: Mouse
Clone: OV-TL 12/30
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: OTN II ovarian carcinoma cell line
Molecular Weight: 54 kDa
Positive Control: Ovarian CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cytokeratin 7 Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Cytoskeletal Protein | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
OV-TL 12/30 | |
Unconjugated | |
OTN II ovarian carcinoma cell line | |
IVD | |
Primary | |
Concentrated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction