Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.

$416.00 - $706.50

Specifications

Antigen Cytokeratin 7 Ab-2
Clone OV-TL 12/30
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 8
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1352P
View Documents
Thermo Scientific™
MS1352P
Purified Ab with BSA and Azide 1mL
Each for $706.50
Add to cart
 
MS1352P1
View Documents
Thermo Scientific™
MS1352P1
Purified Ab with BSA and Azide 500μL
Each for $416.00
Add to cart
 
MS1352P1ABX
View Documents
Thermo Scientific™
MS1352P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $465.00
Add to cart
 
MS1352PABX
View Documents
Thermo Scientific™
MS1352PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $599.00
Add to cart
 
MS1352RQ
View Documents
Thermo Scientific™
MS1352RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $579.50
Add to cart
 
MS1352R1
View Documents
Thermo Scientific™
MS1352R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS1352R7
View Documents
Thermo Scientific™
MS1352R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1352P0
View Documents
Thermo Scientific™
MS1352P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.

Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).

Host Species: Mouse

Clone: OV-TL 12/30

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: OTN II ovarian carcinoma cell line

Molecular Weight: 54 kDa

Positive Control: Ovarian CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Cytokeratin 7 Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
OV-TL 12/30
Monoclonal
Murine
Cytoskeletal Protein
OTN II ovarian carcinoma cell line
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit