Learn More
Cytokeratin 7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168571AP
Catalog No. 50-173-1224
Add to cart
Description
Description
This antibody is specifically against KRT7.
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications
Specifications
Cytokeratin 7
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08729, Q9DCV7
CK 7, CK7, Cytokeratin 7, K2C7, K7, keratin 7, KRT7, Sarcolectin, SCL, Type II keratin Kb7
Rabbit
IgG
150 μL
-20°C
Primary
110310, 3855
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
KRT7
Liquid
KRT7
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse