Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 8/18 Mouse anti-Human, PE/Cy5.5, Clone: KRT8.18/1346, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254520PECY55

Catalog No. N254520PEC5


Add to cart

Description

Description

Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytokeratin 8/18
KRT8.18/1346
Flow Cytometry
Conjugated, Purified
KRT8
Cytoskeleton preparation from HeLa cells
Protein A or G purified
Cancer, Cytoskeleton Markers, Phospho Specific
Primary
3856.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit