Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 8/18 Rabbit anti-Human, PE, Clone: KRT8.18/6579R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314099PE
This item is not returnable.
View return policy
Description
Cytokeratin 8/18 Monoclonal antibody specifically detects Cytokeratin 8/18 in Human samples. It is validated for Flow CytometrySpecifications
Cytokeratin 8/18 | |
Monoclonal | |
PE | |
CARD2, CK8, CK-8, CYK8cytokeratin 8, Cytokeratin-8, K2C8, K8cytokeratin-8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, Type-II keratin Kb8 | |
Recombinant full-length human Cytokeratin 8 and Cytokeratin 18 proteins | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers | |
3856 | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry | |
KRT8.18/6579R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title