Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Cytokeratin 8, Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 8, Rabbit Monoclonal Antibody.

$419.00 - $1910.00

Specifications

Antigen Cytokeratin 8
Clone EP1628Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RM2107R7
View Documents
Thermo Scientific™
RM2107R7
Ready-to-use for IHC (UVLP) 7mL
Each for $419.00
Add to cart
 
RM2107RQ
View Documents
Thermo Scientific™
RM2107RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $523.00
Add to cart
 
RM2107S
View Documents
Thermo Scientific™
RM2107S
Culture Supernatant with Azide 1mL
Each for $1,910.00
Add to cart
 
RM2107S0
View Documents
Thermo Scientific™
RM2107S0
Culture Supernatant with Azide 100μL
Each for $423.50
Add to cart
 
RM2107R1
View Documents
Thermo Scientific™
RM2107R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 8, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Cytokeratin 8 belongs to the type B (basic) subfamily of low molecular weight keratins and usually exists in combination with cytokeratin 18. Cytokeratin 8 is primarily found in the non-squamous simple epithelia. It is expressed in mesothelioma and most adenocarcinomas of simple epithelium including breast ducts, gall bladder, intestine, liver, pancreas and prostate.

Host Species: Rabbit

Clone: EP1628Y

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues on the C-terminus of human CK8

Molecular Weight: 52 kDa

Positive Control: Pancreas, Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Cytokeratin 8
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
EP1628Y
Monoclonal
Rabbit
Cancer and Tumor Biology
A synthetic peptide corresponding to residues of the human CK18
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit