Learn More
Cytokeratin, Acidic (Type I or LMW) Mouse anti-Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit, DyLight 594, Clone: AE1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262221DL594
Catalog No. NB002757
Add to cart
Description
Description
Cytokeratin, Acidic (Type I or LMW) Monoclonal specifically detects Cytokeratin, Acidic (Type I or LMW) in Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
Cytokeratin, Acidic (Type I or LMW)
AE1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry
Purified, Conjugated
Human epidermal keratins
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat, Canine, Chicken, Feline, Primate, Rabbit
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 594
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.1 mL
Primary
The antibody MEM-181 reacts with CD25 (Interleukin-2 receptor alpha chain), a 55 kDa type I transmembrane glycoprotein expressed on activated B and T lymphocytes, activated monocytes/macrophages and on CD4+ T lymphocytes (T regulatory cells); it is lost on resting B and T lymphocytes.